Key Documents
BAF187453011
No-Wire Multi-Tube Racks
number of holes 35
, Holds 4 of 50 mL, 6 of 15 mL, and 25 of 1.5 mL Centrifuge Tubes
About This Item
UNSPSC Code:
41122804
NACRES:
NB.22
Pricing and availability is not currently available.
material
white polypropylene
feature
autoclavable
Holds 25 x 1.5 mL tubes
Holds 4 x 50 mL tubes
Holds 6 x 15 ml tubes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18745-3011
General description
Each No-Wire Racks holds two or three different sizes of round or conical bottom centrifuge tubes. One rack holds four 50ml, six 15ml and twenty-five 1.5ml tubes (hole sizes, 30.2mm, 17.3mm and 11.4mm respectively), other rack holds six 50ml and twelve 15ml centrifuge tubes (holes size, 30.2mm and 17.3mm respectively). Easy to assemble. Dimensions are 246mm x 104mm x 64mm high (9.7" x 4.1" x 2.5"). The white polypropylene racks are submersible and steam autoclavable to 121ºC (250º F).
Store different size tubes of required reagents using one space saving rack
  • Ships flat - Easy to assemble
Legal Information
No-Wire is a trademark of SP Scienceware, Division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)