Documents
Z764035
MicroTissues® 3D Petri Dish® micro-mold toroids
makes toroid-shaped microtissues, fits 12 well plates, 6 x 6 array
Synonym(s):
3D, 3D Cell Culture
About This Item
UNSPSC Code:
41121812
NACRES:
NB.14
material
spherical
sterility
sterile; autoclaved
feature
lid: no
6 x 6 array
packaging
pack of 6 ea
manufacturer/tradename
MicroTissues Inc. 12-36TO
General description
Six autoclavable precision micro-molds to cast 3D Petri Dish for forming toroid shaped microtissues. 3D Petri Dish for use in 12-well plate. Each micro-mold forms 36 toroid shaped recesses in a 6 x 6 array.
  • Nominal dimensions of each toroid shaped 3D culture recess: O.D. 1400 μm x I.D. 600 μm
  • Trough W 400 μm x D 750 μm
  • Micro-molds also form single chamber for cell seeding
When the gelled agarose is removed from the micro-mold, it is transferred to a standard 12 well or 24 well tissue culture dish and equilibrated with cell culture medium. Since the agarose is transparent, the spheroids or microtissues that form at the bottom of each agarose micro-well can be easily viewed using a standard inverted microscope using phase contrast, bright field or fluorescence microscopy.
  • The micro-molds are reusable up to 12 times and can be sterilized via a standard steam autoclave (30 min, dry cycle)
  • The micro-molds should be stored in a covered container to maintain sterility and to avoid collecting dust or fibres on their small features
  • Any microscope with sufficient magnification and resolution functions for cell based views or images will be suitable for use with the 3D Petri Dish products
Legal Information
3D Petri Dish is a registered trademark of MicroTissues Inc.
MicroTissues is a registered trademark of MicroTissues Inc.