Skip to Content
MilliporeSigma

CLS4520

Corning® spheroid microplates

96 well, round black/clear bottom, ULA (Ultra-Low Attachment surface), lid, sterile

Synonym(s):

Corning Spheroid Microplate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.24
UNSPSC Code:
41121800
Material:
clear bottom, polystyrene , round black/clear bottom
Size:
96 wells
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

clear bottom, polystyrene , round black/clear bottom

sterility

sterile

feature

lid, skirt, Specific plates for 3D cell culture, ULA (Ultra-Low Attachment surface)

packaging

pkg of 10 ea, pkg of 50 ea

manufacturer/tradename

Corning 4520

size

96 wells

well volume

300 μL

wells

96 wells

working volume

0.075-0.30 μL

suitability

suitable for (cell cuture)

binding type

(Ultra-Low Attachment)

General description

Key Features:
  • Optically clear round bottom with black opaque microplate body
  • Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
  • Novel proprietary well geometry aids formation of spheroids in center of well
  • Unique design that shields each well to minimize well-to-well cross talk

Key Benefits:
  • Uniform, single spheroid formation across all wells
  • Centering of the spheroid in each well enables automated visualization
  • Culture and assay spheroids in the same microplate, without the need for transfer to a new plate
  • Ease of media exchange or treatment with test compounds/drugs

Application

Corning® spheroid microplates suitable for use in 3D cell culture and uniform spheroid formation with Ultra-Low Attachment surface.

Legal Information

Corning is a registered trademark of Corning, Inc.


Still not finding the right product?

Explore all of our products under Corning® spheroid microplates




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library