Key Documents
Z756709
Scienceware® 50 mL tube holder
grip style, round blue polystyrene
Synonym(s):
centrifuge tube holder, tube rack
About This Item
UNSPSC Code:
41122804
NACRES:
NB.51
material
round blue polystyrene
feature
Holds 1 x 50 mL tube
packaging
bag of 5 ea
manufacturer/tradename
Bel-Art F18795-0001
General description
Leave both hands free for pipetting and other critical tasks. The Grip Style gently grips the tube and moves with the tube when it is lifted from the bench top.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)