Key Documents
Z744000
Worthington Accessories
rack for 10 drawers for LS6000 liquid nitrogen refrigerator
About This Item
UNSPSC Code:
41122800
NACRES:
NB.51
packaging
pack of 1 ea
manufacturer/tradename
Worthington RS60-9C50
technique(s)
cryopreservation: suitable
General description
All Worthington cryostorage refrigerators and dewars each comes with a standard lid, so it is not necessary to purchase an additional one. However, many customers buy spare caps/necktube cores in order to modify them so that different kinds of sample storage devices can be suspended into a storage cannister.