Key Documents
Z740395
The SpecTainer II
size 60 mL, non-sterile
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material
polyethylene cap
polypropylene container
sterility
non-sterile
packaging
pack of 500 ea
manufacturer/tradename
Simport C567-60Y
size
60 mL
General description
For applications needing the utmost security where sample integrity is of high importance. A great feature of the Spectainer I is that it incorporates a unique tamper evident screw cap ensuring peace of mind during transport or storage situations where someone might have tampered with the specimen.
Both containers and caps are manufactured without the use of plasticisers or mold release agents. All material used is free from latex. Containers are 95kpa compliant. Available in three sizes: 60, 90 and 120 ml. The 60 ml size however is not graduated. The sterile container protects its sterility with a tamper evident seal.
Legal Information
SpecTainer is a trademark of Simport