Key Documents
Z648191
STATMARK permanent marker for cassette & slide marking
STATMARK permanent marker for cassette & slide marking
pkg of 12 ea
Synonym(s):
solvent resistant pen
About This Item
UNSPSC Code:
41181600
NACRES:
MA.01
Pricing and availability is not currently available.
packaging
pkg of 12 ea
manufacturer/tradename
Ted Pella 22313
General description
The STATMARK Permanent ink pen in an ultra fine tip is ideal for labeling microscope slides and cassettes. Made of rigid nylon that allow for precise hand marking on glass slides as well as on most tissue embedding cassettes.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1
1 of 1
Questions
1–4 of 4 Questions  
  1. Is this marker solvent resistant to chloroform or low concentration acid, like 10% acetic acid?
    1 answer
    1. When allowed to dry, the marker is resistant chemicals and solvents. Some fading may occur with exposure to ethanol or ethanol/xylene mixtures. See the statement below from the manufacturer.
      Helpful?
  2. This marker writing, is stable up to 800 degree centigrade.
    1 answer
    1. This marker has not been tested in high temperature. There is no heat resistant marker product currently available.
      Helpful?
  3. Is this Statmark marker suitable for labeling tubes with a storage temperature of -80°C?
    1 answer
    1. This marker is not rated for cryogenic temperatures. See the link below to the Nalgene Cyroware marker set, which are suitable for ultra-low temperatures: https://www.sigmaaldrich.com/product/sigma/m6163.
      Helpful?
  4. is this marker solvent resistant (including formalin, xylene, and alcohols)?
    1 answer
    1. The STATMARK permanent marker contains quick drying ink is highly resistant to aqueous based stains, alcohols, xylene and xylene substitutes.
      Helpful?
Reviews
Active Filters
  1. 1 Ratings-Only Review