Key Documents
Z740414
SecurTainer III specimen containers
size 60 mL
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material
polyethylene cap
polypropylene container
packaging
pack of 500 ea
manufacturer/tradename
Simport C577-60W
size
60 mL
General description
Chemically resistant and shatterproof, The SecurTainer III Specimen Containers are supplied with a leakproof screw cap, particularly important when transporting hazardous material. Ideal for transport and storage of urine, sputum and most liquids or particulate samples.
Containers are uniquely stackable, shatter resistant and are manufactured from virgin, high-clarity polypropylene. Available in many sizes from 20 to 120 ml. Containers and caps packaged separately in bags of 100. Non sterile. Functional temperature range: -90 °C to +100 °C.
Legal Information
SecurTainer is a trademark of Simport
Choose from one of the most recent versions:
Certificates of Analysis (COA)