Documents
Z369802
Sigma-Aldrich
Stat-Matic plate washer
8-port Manifold
About This Item
UNSPSC Code:
41122102
NACRES:
NB.22
material
polypropylene plate
sterility
non-sterile
manufacturer/tradename
Tricontinent 0822
size
8 wells
color
none
General description
  • Polypropolyene
  • Non-sterile
Fast, convenient, accurate filling or washing of microplates from an external reservoir. All inert polypropylene-and-glass construction with a PTFE plunger. Set manifold by inserting spacers for 100, 200, or 250 μL and dispense into eight wells at a time (spacing set for 96 well plates).