Skip to Content
MilliporeSigma

M2781

Multiwell plate washer/dispenser manifolds

12 position, straight

Synonym(s):

Plate Washer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41122102
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

color

none

General description

For use with 96 well multiwell plates
  • Dispense or aspirate media, or wash, a full row or column at a time
  • Luer fitting
  • Stainless steel and PTFE construction for easy cleaning and autoclavability
  • Optional repeat kit provides continuous operation capability


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library