Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z369799

Sigma-Aldrich

Stat-Matic plate washer

For Multiwell Plates

Synonym(s):

Plate Washer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.22

material

polypropylene plate

sterility

non-sterile

manufacturer/tradename

Tricontinent 0982

size

8 wells

color

none

Looking for similar products? Visit Product Comparison Guide

General description

Can fill 8 wells at once with 100 μL, 200 μL or 250 μL
Unit includes inlet and outlet tubing, three threaded adapters for common sizes of narrow-mouth reservoir bottles, one 8-port manifold, and a manifold holder.
Fast, convenient, accurate filling or washing of microplates from an external reservoir. All inert polypropylene-and-glass construction with a PTFE plunger. Set manifold by inserting spacers for 100, 200, or 250 μL and dispense into eight wells at a time (spacing set for 96 well plates).

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service