Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z369659

Sigma-Aldrich

SealPlate® film

non-sterile

Synonym(s):

ELISA plate seals, qPCR plate seal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.22

material

polyester film

sterility

non-sterile

feature

piercable: no

packaging

pack of 100 ea

manufacturer/tradename

Excel Scientific 100-SEAL-PLT

parameter

-40 to +120 °C temperature

technique(s)

ELISA: suitable

L

146 mm

W × L

7.94 cm × 14.6 cm
79.4 mm × 141 mm

W

79.4 mm

size

Up to 1536 wells

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

Sheets pre-cut to fit standard multiwell plates. Both film and adhesive are inert and compatible with microplate procedures. Adhesive forms a tight, waterproof seal, preventing cross-contamination and evaporation. SealPlate film is 2 mil (0.05 mm) polyester, with a functional temperature range of -40 to +120 °C.

Application

SealPlate® film is suitable for sealing tissue culture plates for temporary storage, incubation, and containment of biohazards. It can also be used in sensitive enzyme-linked immunosorbent assay (ELISA) for sealing of wells to avoid edge effect.

Legal Information

SealPlate is a registered trademark of Excel Scientific, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about Supel™ BioSPME C18 high-throughput devices to determine plasma protein binding and decrease the sample preparation time to less than an hour.

Learn about Supel™ BioSPME C18 high-throughput devices to determine plasma protein binding and decrease the sample preparation time to less than an hour.

Learn about Supel™ BioSPME C18 high-throughput devices to determine plasma protein binding and decrease the sample preparation time to less than an hour.

Learn about Supel™ BioSPME C18 high-throughput devices to determine plasma protein binding and decrease the sample preparation time to less than an hour.

See All

Protocols

GenomePlex is a Whole Genome Amplification (WGA) method that allows the researcher to generate a representative, 500-fold amplification of genomic DNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service