Documents
AXYPCRSP
AxySeal sealing film with acrylic adhesive, for ELISA assays and T/C plates
size 80 μm, polyester, pkg of 5x100sheets/cs
Synonym(s):
96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate
About This Item
UNSPSC Code:
41122410
NACRES:
NB.22
material
polyester
sterility
non-sterile
manufacturer/tradename
Axygen Inc PCR-SP
Corning
packaging
pkg of 5x100sheets/cs
size
80 μm
General description
  • Axygen® AxySeal sealing films are ideal for sealing tissue culture plates, for short-term storage and incubation, transport, and containment biohazardous solutions.
  • Polyester film with uniform adhesive properties enables efficient well sealing for sensitive ELISA assays
  • The versatile sealing film can be cost-effective in various applications, including PCR and real-time PCR to ELISA and cell culture.
  • Compatible with a wide range of temperatures.
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.