Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A2566

Sigma-Aldrich

Nalgene® centrifuge bottle adapter

LDPE adapter

Synonym(s):

Nalgene® centrifuge bottles, Style 3122

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103911
NACRES:
NB.11

material

translucent polycarbonate
translucent polypropylene screw closure

feature

For use with 250 mL tubes (style 3120,3121, 3122, 3127, 3140, and 3141)

packaging

pack of 6 ea

manufacturer/tradename

Nalgene® DS3125-0250

technique(s)

centrifugation: suitable

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • For use with Nalgene 250ml centrifuge bottles catalog numbers 3120-0250, 3121-0250, 3122-0250, 3127-0250, 3140-0250 and 3141-0250
  • Adapts Nalgene 250ml centrifuge bottles to fit in Thermo Scientific Sorval GSA rotors
  • Molded of white, low-density polyethylene

Other Notes

For all 250-ml centrifuge bottles used in Sorvall six-place GSA rotors.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service