Documents
CLS431842
Corning® centrifuge bottle
with silicon O-ring screw cap, capacity 250 mL, polycarbonate
Synonym(s):
centrifugation bottles, digestion bottles, shaking bottles
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
material
clear bottle
flat bottom tube
polycarbonate
round bottle
wide-mouth bottle
sterility
non-sterile
feature
autoclavable
cap
packaging
pkg of 36 ea
manufacturer/tradename
Corning 431842
parameter
27500 × g max. RCF
capacity
250 mL
Features and Benefits
  • Corning® centrifuge bottles have a specially designed cap for improved grip
  • Includes clear graduations and textured marking spot.
  • Bottles are designed with extra-thick walls for high durability
  • Wide mouth for easier liquid filling
  • Compatible with most centrifuges and do not require special buckets
  • Unique design aid researchers to reduce the number of centrifugation cycles and enhance productivity
Legal Information
Corning is a registered trademark of Corning, Inc.