Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WH0011093M6

Sigma-Aldrich

Monoclonal Anti-ADAMTS13 antibody produced in mouse

clone 4F12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ADAM metallopeptidase with thrombospondin type 1 motif, 13, Anti-C9orf8, Anti-DKFZp434C2322, Anti-MGC118899, Anti-MGC118900, Anti-TTP, Anti-VWFCP, Anti-vWFCP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4F12, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2bκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Related Categories

General description

The gene encoding ADAMTS13 is located on human chromosome 9q34.2 and has 29 exons. It is synthesized by the liver.
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motif) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene is the von Willebrand Factor (vWF)-cleaving protease, which is responsible for cleaving at the site of Tyr842-Met843 of the vWF molecule. A deficiency of this enzyme is associated with thrombotic thrombocytopenic purpura. Alternative splicing of this gene generates multiple transcript variants encoding different isoforms. (provided by RefSeq)

Immunogen

ADAMTS13 (NP_620594, 1328 a.a. ~ 1427 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
FINVAPHARIAIHALATNMGAGTEGANASYILIRDTHSLRTTAFHGQQVLYWESESSQAEMEFSEGFLKAQASLRGQYWTLQSWVPEMQDPQSWKGKEGT

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ADAMTS13 activity and genetic mutations in Japan
Miyata T, et al.
Hamostaseologie, 33(02), 131-137 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service