Skip to Content
MilliporeSigma
All Photos(1)

Documents

TL11430

Sigma-Aldrich

DMT-locT Phosphoramidite

configured for ABI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H49N4O9P
CAS Number:
Molecular Weight:
772.82
UNSPSC Code:
41116105
NACRES:
NA.51

biological source

non-animal source (BSE/TSE no Risk)

Quality Level

product line

Proligo Reagents

Assay

≥98.0% (31P-NMR)
≥98.0% (reversed phase HPLC)

form

powder

impurities

≤3 wt. % Residual Solvent Content
<0.4% Water Content (Karl Fischer)
<0.5% Single unspecified Impurity (reversed phase HPLC)

color

white to off-white

solubility

soluble, clear, colorless

absorption

<0.1 in acetonitrile at 0.2 M

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

compatibility

configured for ABI

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service