Skip to Content
MilliporeSigma

CL11430

DMT-locMeC(bz) Phosphoramidite

configured for ABI

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C48H54N5O9P
CAS Number:
Molecular Weight:
875.94
MDL number:
NACRES:
NA.51
UNSPSC Code:
41116105
Assay:
≥98.0% (31P-NMR), ≥98.0% (reversed phase HPLC)
Biological source:
non-animal source (BSE/TSE no Risk)
Form:
powder
Solubility:
acetonitrile: 0.2 M, clear, colorless
Storage temp.:
−20°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

non-animal source (BSE/TSE no Risk)

Quality Segment

product line

Proligo Reagents

assay

≥98.0% (31P-NMR), ≥98.0% (reversed phase HPLC)

form

powder

impurities

≤3 wt. % Residual Solvent Content, <0.4% Water Content (Karl Fischer), <0.5% Single unspecified Impurity (reversed phase HPLC)

color

white to off-white

solubility

acetonitrile: 0.2 M, clear, colorless

absorption

<0.1 in acetonitrile at 0.2 M

suitability

conforms to structure for H-NMR, conforms to structure for LC-MS

compatibility

configured for ABI

storage temp.

−20°C

General description

DMT-locMeC(bz) Phosphoramidite belongs to the group of LNA phosphoramidite. It is known as DMT-5-Me-locCytidine(N4-Benzoyl)- ß-Cyanoethylphosphoramidite. The synthesis cycle for Locked Nucleic Acid (LNA) oligonucleotides consists of the same series of reactions as the cycle that is employed for DNA monomers. However, the rate of coupling for LNA monomers is slower compared to that of DNA monomers (a coupling time of 8 minutes is recommended for LNA monomers compared to 90 seconds for DNA monomers). With the exception of the LNA monomers, LNA synthesis is accomplished with the same reagents as DNA synthesis.

Features and Benefits

Key Features of LNA Phosphoramidites:

  • LNA oligonucleotides are prepared by phosphoramidite chemistry
  • Standard DNA synthesizer platforms can be employed. No change is required in the reagents commonly used for DNA synthesis.
  • To further enhance the hybridization characteristics of LNA, 5-methyl-cytidine is employed instead of cytidine.
  • LNA monomers are as soluble in acetonitrile as their DNA counterparts (except for the 5-methyl-cytidine derivative, which requires the application of 10-20%, dichloromethane as a co-solvent)
  • Mixmer oligonucleotides containing LNA, DNA and/or RNA monomers can be assembled easily
  • LNA oligonucleotides with predefined melting temperatures (Tm) can be designed and prepared.

Other Notes

LNA phosphoramidites from Merck are diluted with dryacetonitrile, except for locMeC(bz)-phosphoramidite.This phosphoramidite requires the application of a cosolventto prevent crystallization from the solution onthe synthesizer. Dichloromethane or tetrahydrofuran(THF) can be applied as co-solvents with acetonitrile tocompletely dissolve locMeC(bz)-phosphoramidite.

Legal Information



Still not finding the right product?

Explore all of our products under DMT-locMeC(bz) Phosphoramidite


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library