Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T8793

Sigma-Aldrich

Tris Buffered Saline

powder, pH 8.0, with 3% nonfat Milk

Synonym(s):

NFDM blocking buffer, TBS with 3% NFDM, Tris Buffered Saline with 3% Non Fat Dry Milk

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105319
NACRES:
NA.25

product name

Tris Buffered Saline, pH 8.0, with 3% nonfat Milk, powder

form

powder

application(s)

diagnostic assay manufacturing

Looking for similar products? Visit Product Comparison Guide

Packaging

Foil pouches

Reconstitution

Contents of one pouch, when dissolved in 100 mL of distilled or deionized water, will yield 0.05 M Tris buffered saline, pH 8.0 at 25°C, containing 3% (w/v) nonfat milk.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carine Lindquist et al.
Journal of lipid research, 58(7), 1362-1373 (2017-05-06)
Hepatic mitochondrial function, APOC-III, and LPL are potential targets for triglyceride (TG)-lowering drugs. After 3 weeks of dietary treatment with the compound 2-(tridec-12-yn-1-ylthio)acetic acid (1-triple TTA), the hepatic mitochondrial FA oxidation increased more than 5-fold in male Wistar rats. Gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service