T9039
Tris Buffered Saline, with Tween™ 20, pH 8.0
powder, pH 8.0, with Tween® 20
Synonym(s):
TBS, TBST
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105319
NACRES:
NA.25
Recommended Products
Looking for similar products? Visit Product Comparison Guide
Application
Tris Buffered Saline, with Tween® 20, pH 8.0 has been used as a wash buffer in western blotting and enzyme linked immunosorbent assay. It has also been used along with 5% milk to block proteins on nitrocellulose in western blotting.
Biochem/physiol Actions
Tris Buffered Saline with Tween 20 is predominantly used as a wash buffer and also in blocking steps of immunoblotting technique.
Packaging
Foil pouches
Reconstitution
Contents of one pouch, when dissolved in one liter of distilled or deionized water, will yield 0.05 M Tris buffered saline (NaCl - 0.138 M; KCl - 0.0027 M); TWEEN® 20 - 0.05%, pH 8.0, at 25 °C.
Legal Information
TWEEN is a registered trademark of Croda International PLC
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Joey P Ting et al.
PloS one, 13(1), e0190850-e0190850 (2018-01-13)
To date, IL-17A antibodies remain the only therapeutic approach to correct the abnormal activation of the IL-17A/IL-17R signaling complex. Why is it that despite the remarkable success of IL-17 antibodies, there is no small molecule antagonist of IL-17A in the
Differential antibody responses to gliadin-derived indigestible peptides in patients with schizophrenia
McLean RT, et al.
Translational Psychiatry, 7(5), e1121-e1121 (2017)
Study of circulating IgG antibodies to BIRC5 and MYC in non-small cell lung cancer
Ye Leiguang, et al.
FEBS Open Bio, 5(1), 809-812 (2015)
Progestin treatment decreases CD133+ cancer stem cell populations in endometrial cancer
Guy MS, et al.
Gynecologic Oncology, 140(3), 518-526 (2016)
Matrix cells from Wharton's jelly form neurons and glia
Mitchell KE, et al.
Stem Cells, 21(1), 50-60 (2003)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service