Skip to Content
MilliporeSigma

C9483

Coating Stabilizer and Blocking Buffer

liquid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.46
UNSPSC Code:
12352203
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

Quality Segment

form

liquid

storage temp.

2-8°C

General description

Contains 24 ppm 5-chloro-2-methyl-4-isothiazolin-3-one and 8 ppm 2-methyl-4-isothiazolin-3-one.

Application

The solution can be used to improve the stability and function of antigens and other very unstable proteins bound to a solid phase. It can be used as supplied for very unstable proteins, or diluted 1:1 with currently used blocking agent for less labile antigens and proteins. Coating Stabilizer and Blocking Buffer has been used as a blocking agent in transamidation assay to evaluate gliadin and other modelled ligands.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

10 - Combustible liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library