Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5020

Sigma-Aldrich

DDR1 (444-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CAK, CD167, DDR, EDDR1, MCK10, NEP, NTRK4, PTK3, PTK3A, RTK6, TRKE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

4.25-5.75 nmol/min·mg

mol wt

~77 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... DDR1(780)

General description

DDR1 is a member of the subfamily of tyrosine kinase receptors with a homology region to the Dictyostelium discoideum protein discoidin I in its extracellular domain. DDR1 is widely expressed in normal and transformed epithelial cells and is activated by various types of collagen. Expression of DDR1 is restricted to epithelial cells, particularly in the kidney, lung, gastrointestinal tract, and brain and DDR1 is over-expressed in fast-growing invasive tumors of the breast, ovary, esophagus, brain and lung. Collagen type I mediated stimulation of DDR1 leads to activation of matrix metalloproteinase 2 and 9 as well as increased cell invasion.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daizo Yoshida et al.
Cell adhesion & migration, 1(2), 92-98 (2007-04-01)
The aim of this study is to examine a novel drop culture model using a biologically inspired self-assembling peptide: hydrogel (RAD16-I, also called PuraMatrix), which produces a nanoscale environment similar to native extracellular matrix (ECM) for a cell line weakly
E Di Marco et al.
The Journal of biological chemistry, 268(32), 24290-24295 (1993-11-15)
We have identified and cloned a new member of the trk gene family, termed trkE, which generates a 3.9-kilobase (kb) transcript in normal human keratinocytes and in a variety of normal human tissues, but not in liver. Albeit at low

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service