Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

158127

Sigma-Aldrich

Paraformaldehyde

powder, 95%

Synonym(s):

Polyoxymethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2O)nH
CAS Number:
Molecular Weight:
30.03 (as monomer)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

1.03 (vs air)

Quality Level

vapor pressure

<1.45 mmHg ( 25 °C)

Assay

95%

form

powder

autoignition temp.

572 °F

expl. lim.

73 %

mp

120-170 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[O-]#[C+H2]

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Paraformaldehyde is also referred as polyoxymethylene. It participates as an external CO source in the synthesis of aromatic aldehydes and esters.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Flam. Sol. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morphological and physiological species-dependent characteristics of the rodent Grueneberg ganglion
Brechbuhl J, et al.
Frontiers in Neuroanatomy, 8, 87-87 (2014)
The effects of simvastatin on hippocampal caspase-3 and Bcl-2 expression following kainate-induced seizures in rats
Sun J, et al.
International Journal of Molecular Medicine, 30(4), 739-746 (2012)
Flow acetone-staining technique: A highly efficient procedure for the simultaneous analysis of DNA content, cell morphology, and immunophenotype by flow cytometry
Carbonari M, et al.
Cytometry. Part A : the Journal of the International Society For Analytical Cytology, 73(2), 168-174 (2008)
In Situ Hybridization in Neurobiology, 27-27 (1994)
Cytometry, 256-256 (2000)

Protocols

Formaldehyde cross-linking protocol detects specific protein interactions with confidence.

Formaldehyde cross-linking protocol detects specific protein interactions with confidence.

Formaldehyde cross-linking protocol detects specific protein interactions with confidence.

Formaldehyde cross-linking protocol detects specific protein interactions with confidence.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service