Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4850

Sigma-Aldrich

Heat Shock Protein 22 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

α-crystallin C chain, E2IG1, HMN2A, HSP22, HSPB8 H11 HMN2 CMT2L DHMN2, Heat shock protein β-8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

~22 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HSPB8(26353)

General description

Recombinant Human Heat Shock Protein is produced in E. coli and purified by proprietary chromatographic techniques.

Physical form

Lyophilized powder.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in H20 to a concentration of 0.1-1.0 mg/mL. The solution can then be diluted to other aqueous buffers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service