Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0183

Sigma-Aldrich

Hsp90b Active human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

HSP84, HSP90Beta, HSPC2, Heat shock protein 90 kDa β

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥80% (SDS-PAGE)

form

aqueous solution

mol wt

83 kDa

packaging

pkg of 200 μg

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HSP90AB1(3326)

General description

Hsp90b (heat shock protein 90-β) is a constitutive cytosolic isoform of Hsp90. The gene is mapped to human chromosome 6p21.1. Hsp90b is an ATP-dependent molecular chaperone and is upregulated in many tumor cells.
Human Heat Shock Protein 90β (GenBank Accession No. AY359878), full length with C-terminal His tag, MW = 83kDa, expressed in an Escherichia coli expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Biochem/physiol Actions

Hsp90 (heat shock protein 90) is needed for folding, maturation, transport, and degradation of various proteins, particularly signaling molecules, steroid receptors, and transcription factors. Hsp90b (Hsp90 β) is secreted by various cells and works as a chaperone for selected surface-associated or secreted proteins. It acts as a chaperone for matrix metalloproteinases and extracellular matrix proteins, and helps in cell motility. Suppression of the Hsp90b activity is suggested as a therapeutic approach for various cancers and inflammatory diseases.

Physical form

Formulated in 25 mM Tris-HCl, pH 7.5, 100mM NaCl, 0.05% Tween 20, 20% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Hsp90AB1 Protein is Overexpressed in Non-small Cell Lung Cancer Tissues ?and Associated with Poor Prognosis in Lung Adenocarcinoma Patients].
Wang M, et al.
Zhongguo Fei ai za Zhi = Chinese Journal of Lung Cancer, 19, 64-64 (2016)
Cell surface heparan sulfate proteoglycans are involved in the binding of Hsp90a and Hsp90? to the cell plasma membrane.
Snigireva AV, et al.
Cell adhesion & migration, 9, 460-460 (2015)
HSP90AB1: Helping the good and the bad.
Haase M, et al.
Gene, 575, 171-171 (2016)
DNA copy number analysis of Grade II-III and Grade IV gliomas reveals differences in molecular ontogeny including chromothripsis associated with IDH mutation status.
Cohen A, et al.
Acta Neuropathologica Communications, 3, 34-34 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service