Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H7533

Sigma-Aldrich

Heat Shock Protein 32 from rat

recombinant, expressed in E. coli, buffered aqueous solution, ≥90% (SDS-GE)

Synonym(s):

HSP 32, Heme oxygenase-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

rat

Quality Level

recombinant

expressed in E. coli

Assay

≥90% (SDS-GE)

form

buffered aqueous solution

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

Physical form

Solution in 50 mM ammonium bicarbonate, pH 8.3

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Shibahara et al.
Proceedings of the National Academy of Sciences of the United States of America, 82(23), 7865-7869 (1985-12-01)
Two cDNA clones for rat heme oxygenase have been isolated from a rat spleen cDNA library in lambda gt11 by immunological screening using a specific polyclonal antibody. One of these clones has an insert of 1530 nucleotides that contains the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service