Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP3271

Sigma-Aldrich

GM-CSF rat

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

CSF-3, GM-CSF?, MGI-1G, pluripoietin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.75

biological source

rat

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≥0.01 ng/mL

mol wt

14.5 kDa

packaging

pkg of 20 μg

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... CSF2(12981)

General description

It is produced in by endothelial cells, monocytes, fibroblasts and T-lymphocytes. GM-CSF inhibits neutrophil migration and enhances the functional activity of the mature end-cells. The human and murine molecules are species-specific and exhibit no cross-species reactivity.

Biochem/physiol Actions

GM-CSF is a hematopoietic growth factor that stimulates the development of neutrophils and macrophages and promotes the proliferation and development of early erythroid megakaryocytic and eosinophilic progenitor cells. Recombinant murine GM-CSF is a 14.5 kDa globular protein consisting of 128 amino acids residues.

Physical form

Lyophilized from 10 mM Tris, pH 8.5

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Nemunaitis et al.
Blood, 77(9), 2065-2071 (1991-05-01)
Forty-seven patients with hematologic neoplasia received recombinant human granulocyte-macrophage colony-stimulating factor (rhGM-CSF) by daily 2-hour infusion following allogeneic bone marrow transplantation from HLA-identical sibling donors in a phase I-II dose-escalation trial. Dose levels ranged from 30 to 500 micrograms/m2/d. At

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service