Skip to Content
MilliporeSigma
All Photos(2)

Documents

SRP0354

Sigma-Aldrich

Aurora Kinase A active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥90% (SDS-PAGE)

Synonym(s):

ARK1, AURA, BTAK, STK6

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

50 kDa

packaging

pkg of 10 μg

technique(s)

activity assay: suitable
inhibition assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... AURKA(6790)

General description

AURA/AURKA (aurora kinase A) is mapped to human chromosome 20q13.2. It is also termed as STK15 (serine/threonine kinase 15). This gene codes for a protein of 403 amino acids with regulatory and catalytic domains. AURA is overexpressed in several malignancies like invasive carcinomas of ovary, head, neck and breast.
Human Aurora kinase A (GenBank Accession No. NM_003600), amino acids 2- 403 with N-terminal HIS6-tag, MW=50 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Biochem/physiol Actions

AURA/AURKA (aurora kinase A) modulates cell cycle checkpoints and plays important roles in centrosome division, cytoskeleton and cytokinesis. It sustains the integrity of chromosomes after DNA damage. The catalytic domain of AURKA possesses a serine/threonine kinase activity.

Unit Definition

One unit is defined as the amount of enzyme that will phosphorylate 1 pmol of Ser/Thr 1 peptide substarte per minute at pH 7.4 and 30°C.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A single nucleotide polymorphism in codon F31I and V57I of the AURKA gene in invasive ductal breast carcinoma in Middle East
Golmohammadi R, et al.
Medicine, 96(37) (2017)
Mutsuko Ouchi et al.
The Journal of biological chemistry, 279(19), 19643-19648 (2004-03-03)
Aurora-A/BTAK/STK15 localizes to the centrosome in the G(2)-M phase, and its kinase activity regulates the G(2) to M transition of the cell cycle. Previous studies have shown that the BRCA1 breast cancer tumor suppressor also localizes to the centrosome and
Naoko Kunitoku et al.
Developmental cell, 5(6), 853-864 (2003-12-12)
The Aurora (Ipl1)-related kinases are universal regulators of mitosis. We now show that Aurora-A, in addition to Aurora-B, regulates kinetochore function in human cells. A two-hybrid screen identified the kinetochore component CENP-A as a protein that interacts with Aurora-A. Aurora-A
Sunyi Lee et al.
FEBS letters, 588(17), 2844-2850 (2014-07-06)
IK is known to inhibit the expression of major histocompatibility complex (MHC) class II antigen, but other cellular functions of IK remain to be uncovered. In this study, IK depletion caused misalignment of chromosomes through an increase in Aurora A
Veronica Rendo et al.
Nature communications, 11(1), 1308-1308 (2020-03-13)
Cancer chemotherapy targeting frequent loss of heterozygosity events is an attractive concept, since tumor cells may lack enzymatic activities present in normal constitutional cells. To find exploitable targets, we map prevalent genetic polymorphisms to protein structures and identify 45 nsSNVs (non-synonymous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service