MilliporeSigma
  • CENP-A phosphorylation by Aurora-A in prophase is required for enrichment of Aurora-B at inner centromeres and for kinetochore function.
CENP-A phosphorylation by Aurora-A in prophase is required for enrichment of Aurora-B at inner centromeres and for kinetochore function.
Developmental cell (2003-12-12)
Naoko Kunitoku, Takashi Sasayama, Tomotoshi Marumoto, Dongwei Zhang, Shinobu Honda, Osamu Kobayashi, Katsuyoshi Hatakeyama, Yukitaka Ushio, Hideyuki Saya, Toru Hirota
ABSTRACT
The Aurora (Ipl1)-related kinases are universal regulators of mitosis. We now show that Aurora-A, in addition to Aurora-B, regulates kinetochore function in human cells. A two-hybrid screen identified the kinetochore component CENP-A as a protein that interacts with Aurora-A. Aurora-A phosphorylated CENP-A in vitro on Ser-7, a residue also known to be targeted by Aurora-B. Depletion of Aurora-A or Aurora-B by RNA interference revealed that CENP-A is initially phosphorylated in prophase in a manner dependent on Aurora-A, and that this reaction appears to be required for the subsequent Aurora-B-dependent phosphorylation of CENP-A as well as for the restriction of Aurora-B to the inner centromere in prometaphase. Prevention of CENP-A phosphorylation also led to chromosome misalignment during mitosis as a result of a defect in kinetochore attachment to microtubules. Our observations suggest that phosphorylation of CENP-A on Ser-7 by Aurora-A in prophase is essential for kinetochore function.
Sigma-Aldrich
Aurora Kinase A active human, recombinant, expressed in baculovirus infected Sf9 cells, ≥90% (SDS-PAGE)