Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.04014

Supelco

Glass beads 2 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NB.51

storage temp.

no temp limit

Quality Level

General description

Glass beads can be used to purify, enrich or separate samples of solids, liquids or gases. They offer excellent batch consistency to ensure reliable, reproducible results.

Application

Glass beads 2 mm can be used as a supporting matrix to develop biologically inert and reusable liquid culture media. It is also used as solid support to immobilize metal-doped TiO2 photocatalysts by heat attachment method, to prevent agglomeration and easy separation of doped nanoparticles.

Analysis Note

Identity: conforms

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service