Skip to Content

D5197

SigMatrix Ultra Serum Diluent

Synonym(s):

Serum Diluent

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.25
UNSPSC Code:
12352202
Biological source:
human
Form:
solution
Technique(s):
LC/MS: suitable
Concentration:
18—22 mg/mL protein
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in rice

form

solution

concentration

18—22 mg/mL protein

technique(s)

LC/MS: suitable

shipped in

wet ice

storage temp.

−20°C

General description

For more information on other Sigmatrix products visit the Sigmatrix Selection Guide.

Application

This solution is provided ready-to-use as a 2% (w/v) recombinant HSA (Human Serum Albumin) in PBS (Phosphate Buffered Saline), pH 7.4. This formulation has been shown to be appropriate for use as a sample diluent or matrix for preparing calibrants/controls for clinical diagnostic LC-MS/MS assays. Beneficially, as the HSA source is recombinant, the matrix should not contain any measurable contaminants typically found in even doubly stripped serum. SigMatrix Ultra has been successfully used in LC-MS/MS assays for clinically relevant concentrations of total testosterone (20–2,000 pg/mL), estradiol, and estrone (2–1,000 pg/mL). Calibrators prepared in SigMatrix Ultra showed good linearity (r2 >0.97) and no interference at the lowest calibrator concentrations for all analytes tested. Additionally, extracted blank SigMatrix Ultra spiked with analyte specific isotopicallylabelled internal standards showed no interferences above the lower limits of quantitation at the retention time of the respective analytes.

Physical form

SigMatrix Serum Diluent is an aqueous solution containing 2% highly purified recombinant human albumin (expressed in rice) in phosphate buffered saline (pH 7.4) with no preservatives


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library