Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3907

Sigma-Aldrich

CNS-11g

≥98% (HPLC)

Synonym(s):

2-(4-benzyl-1-oxo-2(1H)-phthalazinyl)-N-(2,6-dimethylphenyl)acetamide, N-(2,6-Dimethylphenyl)-1-oxo-4-(phenylmethyl)-2(1H)-phthalazineacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H23N3O2
CAS Number:
Molecular Weight:
397.47
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

Biochem/physiol Actions

Brain tissue penetrant alpha-synuclein ligand that potently disaggregate alpha-synuclein fibrils in vitro and in vivo models.
CNS-11g is brain tissue penetrant alpha-synuclein ligand that potently disaggregate alpha-synuclein fibrils in vitro and in vivo models. CNS-11g inhibits intracellular seeding of a-synuclein fibrils in biosensor cells-HEK293T cells and mitigate a-synuclein cytotoxicity in Neuro-2a (N2a) neuronal cells. C-11g reduces number of a-synuclein aggregates in C. elegans model.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin A Murray et al.
Proceedings of the National Academy of Sciences of the United States of America, 120(7), e2217835120-e2217835120 (2023-02-10)
The amyloid aggregation of alpha-synuclein within the brain is associated with the pathogenesis of Parkinson's disease (PD) and other related synucleinopathies, including multiple system atrophy (MSA). Alpha-synuclein aggregates are a major therapeutic target for treatment of these diseases. We identify

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service