MilliporeSigma
  • Small molecules disaggregate alpha-synuclein and prevent seeding from patient brain-derived fibrils.
Small molecules disaggregate alpha-synuclein and prevent seeding from patient brain-derived fibrils.
Proceedings of the National Academy of Sciences of the United States of America (2023-02-10)
Kevin A Murray, Carolyn J Hu, Hope Pan, Jiahui Lu, Romany Abskharon, Jeannette T Bowler, Gregory M Rosenberg, Christopher K Williams, Gazmend Elezi, Melinda Balbirnie, Kym F Faull, Harry V Vinters, Paul M Seidler, David S Eisenberg
ABSTRACT
The amyloid aggregation of alpha-synuclein within the brain is associated with the pathogenesis of Parkinson's disease (PD) and other related synucleinopathies, including multiple system atrophy (MSA). Alpha-synuclein aggregates are a major therapeutic target for treatment of these diseases. We identify two small molecules capable of disassembling preformed alpha-synuclein fibrils. The compounds, termed CNS-11 and CNS-11g, disaggregate recombinant alpha-synuclein fibrils in vitro, prevent the intracellular seeded aggregation of alpha-synuclein fibrils, and mitigate alpha-synuclein fibril cytotoxicity in neuronal cells. Furthermore, we demonstrate that both compounds disassemble fibrils extracted from MSA patient brains and prevent their intracellular seeding. They also reduce in vivo alpha-synuclein aggregates in C. elegans. Both compounds also penetrate brain tissue in mice. A molecular dynamics-based computational model suggests the compounds may exert their disaggregating effects on the N terminus of the fibril core. These compounds appear to be promising therapeutic leads for targeting alpha-synuclein for the treatment of synucleinopathies.
Sigma-Aldrich
CNS-11g, ≥98% (HPLC)
Sigma-Aldrich
CNS-11, ≥98% (HPLC)