Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3710

Sigma-Aldrich

Linaclotide

≥95% (HPLC)

Synonym(s):

CCEFCCNPACTGCF-OH cyclo(1-6),(2-10),(5-13)-tris(disulfide), Cys-Cys-Glu-Tyr-Cys-Cys-Asn-Pro-Ala-Cys-Thr-Gly-Cys-Tyr-OH cyclo(1-6),(2-10),(5-13)-tris(disulfide), L-Cysteinyl-L-cysteinyl-L-α-glutamyl-L-tyrosyl-L-cysteinyl-L-cysteinyl-L-asparaginyl-L-prolyl-L-alanyl-L-cysteinyl-L-threonylglycyl-L-cysteinyl-L-tyrosine-OH cyclo(1-6),(2-10),(5-13)-tris(disulfide)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C59H79N15O21S6
CAS Number:
Molecular Weight:
1526.74
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

(Powder or Lyophilized powder or film)

storage condition

desiccated

color

white to off-white

storage temp.

-10 to -25°C

SMILES string

S1SC[C@@H]2NC(=O)[C@@H](NC(=O)[C@H]3N(CCC3)C(=O)[C@@H](NC(=O)[C@H]4NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](CSSC4)N)C1)CCC(=O)O)Cc6ccc(cc6)O)CSSC[C@H](NC(=O)CNC(=O)C(NC2=O)[C@H](O)C)C(=O)N[C@@H](Cc5ccc(cc5)O)C(=O)O)CC(=O)N)C

InChI

1S/C59H79N15O21S6/c1-26-47(82)69-41-25-101-99-22-38-52(87)65-33(13-14-45(80)81)49(84)66-34(16-28-5-9-30(76)10-6-28)50(85)71-40(54(89)72-39(23-97-96-20-32(60)48(83)70-38)53(88)67-35(18-43(61)78)58(93)74-15-3-4-42(74)56(91)63-26)24-100-98-21-37(64-44(79)19-

InChI key

KXGCNMMJRFDFNR-IURAOCFRSA-N

Related Categories

Biochem/physiol Actions

Linaclotide is an orally active guanylate cyclase-C (GC-C) agonist that competes against bacterial STa enterotoxin binding to intestinal mucosal membranes from wild-type, but not GC-C null, mice (Ki = 16.4 nM). Linaclotide stimulates fluid secretion, upregulates cGMP accumulation in the intestinal lumen (5 μg/100 μL via injection into surgically ligated intestinal loops), and significantly increases the rate of gastrointestinal transit when administered orally in mice in vivo (100 μg/kg p.o.).
Orally active guanylate cyclase-C (GC-C) agonist that elicits in vivo pharmacological effects locally in the gastrointestinal tract.

Caution

Hygroscopic

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John Cuppoletti et al.
BMC pharmacology, 12, 3-3 (2012-05-05)
Linaclotide has been proposed as a treatment for the same gastrointestinal indications for which lubiprostone has been approved, chronic idiopathic constipation and irritable bowel syndrome with constipation. Stressors damage the epithelial cell barrier and cellular homeostasis leading to loss of
Alexander P Bryant et al.
Life sciences, 86(19-20), 760-765 (2010-03-24)
Linaclotide is an orally administered 14-amino acid peptide being developed for the treatment of constipation-predominant irritable bowel syndrome (IBS-C) and chronic constipation. We determined the stability of linaclotide in the intestine, measured the oral bioavailability, and investigated whether the pharmacodynamic
Robert W Busby et al.
European journal of pharmacology, 649(1-3), 328-335 (2010-09-25)
Linaclotide is a first-in-class, orally administered 14-amino acid peptide that is in development for the treatment of irritable bowel syndrome with constipation and chronic constipation. We have characterized the solution structure of linaclotide, the in vitro binding and agonist activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service