Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S1-M

Sigma-Aldrich

Human Serum, Normal

Synonym(s):

Normal Human Serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105901
eCl@ss:
26020100
NACRES:
NA.71
biological source:
human

biological source

human

antibody form

serum

manufacturer/tradename

Chemicon®

technique(s)

blocking: suitable (interfering antibodies)

General description

Normal human serum collected off the clot from healthy normal humans. Each donor unit was tested and found negative for HIV 1&2, HIV-Ag, HCV, HBsAg, and RPR by FDA approved methods.

Application

Normal Human Serum has been used as a reference or control in degradation assays involving DNase I and MNase treatments, providing a comparison for assessing the impact of these enzymes on the DHM structures.

Physical form

Nonsterile liquid. Contains no preservatives.

Filtered through a 0.2μ absolute membrane filter before bottling.

Storage and Stability

Store frozen at -20°C for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Midori L Wasielewski et al.
International journal of molecular sciences, 24(4) (2023-02-26)
This study analyzed the nuclease- and serum-driven degradation of millimeter-scale, circular DNA-histone mesostructures (DHMs). DHMs are bioengineered chromatin meshes of defined DNA and histone compositions designed as minimal mimetics of physiological extracellular chromatin structures, such as neutrophil extracellular traps (NETs).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service