Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SML3698

Sigma-Aldrich

G150

≥98% (HPLC)

Synonym(s):

1-(9-(6-Aminopyridin-3-yl)-6,7-dichloro-3,4-dihydro-1H-pyrido[4,3-b]indol-2(5H)-yl)-2-hydroxyethanone, 1-[9-(6-Amino-3-pyridinyl)-6,7-dichloro-1,3,4,5-tetrahydro-2H-pyrido[4,3-b]indol-2-yl]-2-hydroxyethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16Cl2N4O2
CAS Number:
Molecular Weight:
391.25
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

G150 is a substrate site-targeting, potent and selective human cyclic GMP-AMP synthase (cGAS) inhibitor (IC50 = 10.2 nM; 100 nM h-cGAS, 0.1 mM ATP & GTP, 25 nM 100-bp dsDNA) with no murine cGAS potency (IC50 >25 μM) and low cytotoxicity (THP1 LD50 = 53 μM). G150 selectively inhibits dsDNA-induced IFNB1/CXCL10 transcription in THP1 (IC50 = 1.96/7.57 μM) & primary human macrophages (IC50 = 0.62/0.87 μM) without affecting adenylyl cyclase activity, direct STING activation by cGAMP, or immune responses induced by hpRNA (RIG-I pathway), poly(I:C) (OAS/RNase L pathway), LPS (TLR4 pathway).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of human cGAS-specific small-molecule inhibitors for repression of dsDNA-triggered interferon expression
Nature Communications, 10(1), 2261-2261 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service