Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1556

Sigma-Aldrich

AEM1

≥98% (HPLC)

Synonym(s):

ARE expression modulator 1, N-(1,3-Benzodioxol-5-ylmethyl)-5-(4-fluorophenyl)-thieno[2,3-d]pyrimidin-4-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14FN3O2S
CAS Number:
Molecular Weight:
379.41
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)c2c3c([s]c2)[nH]cn[c]3=NCc4cc5c(cc4)OCO5

InChI

1S/C20H14FN3O2S/c21-14-4-2-13(3-5-14)15-9-27-20-18(15)19(23-10-24-20)22-8-12-1-6-16-17(7-12)26-11-25-16/h1-7,9-10H,8,11H2,(H,22,23,24)

InChI key

QGYLTGSHWIGJJD-UHFFFAOYSA-N

Biochem/physiol Actions

AEM1 (ARE expression modulator 1) is also a selective inhibitor of sirtuin 2, which is a protein deacetylase mediated by NAD+. Sirtuin 2 supports tumor cell proliferation, thus its inhibition stimulates cell death in tumors.
AEM1 is a potent inhibitor of deregulated Nrf2 transcriptional activity.
AEM1 is a potent inhibitor of deregulated Nrf2 transcriptional activity. AEM1 broadly decreases the expression of NRF2 controlled genes and sensitizes A549 cells to various chemotherapeutic agents.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

AK-1, a specific SIRT2 inhibitor, induces cell cycle arrest by downregulating Snail in HCT116 human colon carcinoma cells.
Cheon M G, et al.
Cancer Letters, 356(2), 637-645 (2015)
Gesine Hoffmann et al.
The Journal of biological chemistry, 289(8), 5208-5216 (2014-01-01)
Sirtuin 2 (SIRT2) is an NAD(+)-dependent protein deacetylase whose targets include histone H4 lysine 16, p53, and α-tubulin. Because deacetylation of p53 regulates its effect on apoptosis, pharmacological inhibition of SIRT2-dependent p53 deacetylation is of great therapeutic interest for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service