Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SML3699

Sigma-Aldrich

G140

≥98% (HPLC)

Synonym(s):

1-(6,7-Dichloro-9-(1-methyl-1H-pyrazol-3-yl)-3,4-dihydro-1H-pyrido[4,3-b]indol-2(5H)-yl)-2-hydroxyethanone, T015T-010-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16Cl2N4O2
CAS Number:
Molecular Weight:
379.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Substrate site-targeting, non-cytotoxic, potent and selective human cyclic GMP-AMP synthase (cGAS) inhibitor against cellular STING signaling.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of human cGAS-specific small-molecule inhibitors for repression of dsDNA-triggered interferon expression.
Nature Communications, 10(1), 2261-2261 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service