Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3577

Sigma-Aldrich

MMG-11

≥98% (HPLC)

Synonym(s):

Ethyl 5-(2-oxo-2-(2,3,4-trihydroxyphenyl)ethyl)furan-2-carboxylate, MMG 11, MMG11

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14O7
CAS Number:
Molecular Weight:
306.27
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, white to very dark gray

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(OCC)C1=CC=C(O1)CC(C2=CC=C(C(O)=C2O)O)=O

InChI key

IIDUJWIVMGALOG-UHFFFAOYSA-N

Biochem/physiol Actions

MMG-11 is a toll-like receptor 2 (TLR2) antagonist that selectively blocks TLR2/1 over TLR2/6 heterodimer-mediated sginaling in both human & murine species (NF-κB/AP-1 activation IC50 = 0.87 µM vs. 7.4 µM in hTLR2/1 or hTLR2/6 HEK293 reporter cells stimulated with 10 ng/mL TLR2/1 agonist Pam3CSK4 or 1 ng/mL TLR2/6 agonist Pam2CSK4; TNF/IL-6 secretion IC50 = 3.3/0.19 µM against 10 ng/mL Pam3CSK4 vs. 58/14 µM against 1 ng/mL Pam2CSK4 using murine macrophage RAW 264.7). In contrast, the TLR2 antagonist CU-CPT22 is selective for hTLR2/6 over hTLR2/1, while being weakly selective for mTLR2/1 over mTLR2/6 heterodimer.
Toll-like receptor 2 (TLR2) antagonist that selectively blocks TLR2/1 over TLR2/6 heterodimer-mediated sginaling in both human & murine species.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of a pyrogallol derivative as a potent and selective human TLR2 antagonist by structure-based virtual screening
Biochemical Pharmacology, 154, 148-160 (2018)
Maria Grabowski et al.
Biochemical pharmacology, 171, 113687-113687 (2019-11-05)
Toll-like receptor 2 (TLR2) forms heterodimers with either TLR1 or TLR6 to induce protective early inflammatory responses to pathogen- and damage-associated molecular patterns. However, excessive activation is associated with inflammatory and metabolic diseases. Several TLR2 antagonists have been described but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service