Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

UFC5003

Millipore

Amicon® Ultra Centrifugal Filter, 3 kDa MWCO

greener alternative

sample volume 0.5 mL, regenerated cellulose membrane, MWCO 3 kDa

Synonym(s):

Centrifugal concentrator, 3 kDa MWCO, 0.5 mL sample volume

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
eCl@ss:
36100101
NACRES:
NB.24
Pricing and availability is not currently available.

material

polypropylene cap
polypropylene tube
regenerated cellulose membrane
styrene-butadiene housing

Quality Level

sterility

non-sterile

product line

Amicon®

manufacturer/tradename

Amicon® Ultra

sustainability

Greener Alternative Product

parameter

0.5 mL sample volume

technique(s)

protein extraction: suitable
protein purification: suitable

L

4.99 cm (1.96 in.)

diam.

10.8 mm

filtration area

1 cm2

volume

0.5 mL

working volume

0.5 mL

pore size

3 kDa MWCO

greener alternative category

shipped in

ambient

General description

Amicon® Ultra-0.5 centrifugal filter devices provide fast ultrafiltration, with the capability for high concentration factors and easy concentrate recovery from dilute and complex sample matrices. For example, Cytochrome c (12.4 kDa) at 0.25 mg/ml initial concentration is typically retained at >95%.

The vertical design and available membrane surface area provide fast sample processing, high sample recovery (typically greater than 90% of dilute starting solution), and the capability for 30-fold concentration. Solute polarization and subsequent fouling of the membrane are minimized by the vertical design, and a physical deadstop in the filter device prevents spinning to dryness and potential sample loss. Efficient recovery of the concentrated sample (retained species) is achieved by a convenient reverse spin step after collecting the filtrate.

Amicon® Ultra-0.5 devices are supplied non-sterile and are for single use only. With their vertical membrane design and reverse spin, Amicon® Ultra 0.5 mL filters deliver great performance and lower spin times.
We are committed to bringing you Greener Alternative Products, which have been re-engineered to enable more sustainable research solutions. The inner filtration device is made from bio-circular material allocated using the ISCC mass balance approach. Click here for more information.

Application

  • Ultrafiltration with 3,000 Dalton molecular weight cutoff Ultracel® membrane
  • Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
  • Purification of macromolecular components found in tissue culture extracts or cell lysates, primer removal, linkers, or macromolecular labels from a reaction mix, and protein removal prior to HPLC
  • Desalting, buffer exchange, and protein dialysis

Features and Benefits

  • Ultracel® regenerated cellulose membrane with 3 kDa NMWCO delivers retentate recovery of greater than 90%
  • Concentrates 500 μL down to 15 μL
  • Concentration factor 25x to 30x
  • High sample recovery (greater than 90% of dilute starting solution)
  • Fast processing time: 10 to 30 minutes
  • Reverse spin capability provides consistent recoveries

Physical form

Color Code: Clear

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Commercially available protein labeling kits provide researchers with the flexibility to customize their own immunoassay detection panels.

Protein purification is simpler with Amicon Pro. The device delivers consistent sample preparation for more reliable recovery, uncompromised purity and easier data generation.

Filtration methods and membrane filter selection in nanoparticle purification and production.

Filtration methods and membrane filter selection in nanoparticle purification and production.

See All

Protocols

If an antibody must be purified from a small sample volume (0.5-10 mL), then the Amicon® Pro Purification System is recommended.

Detergents are frequently used to solubilize proteins and nucleic acids during purification, but the presence of detergents may interfere with downstream analyses.

Questions

1–7 of 7 Questions  
  1. What is the margin of error for the MWCO? I read somewhere the RECOMMENDATION is to use a MWCO 2-3x smaller than your target. But I was wondering what’s the standard deviation from 3 kDa?

    1 answer
    1. For most membranes, the nominal molecular weight limit, also referred to as the molecular weight cut-off, is determined experimentally under a standard set of operating conditions. These analyses typically employ purified globular proteins to serve as markers or indicators of the retention characteristics of an ultrafiltration membrane. An ultrafiltration membrane with a stated nominal molecular weight limit should retain (reject) at least 90% of a globular solute of that molecular weight in daltons. However, for a wider safety margin, the selected cut-off should be well below the molecular weight of the solute to be retained. It is recommended to use a cut-off 2-3 times smaller than the molecular weight of the protein to be concentrated.

      Helpful?

  2. Is this product compatible with organic solvents, such as DMSO or DMF?

    1 answer
    1. The Amicon Ultra products are suitable for use with samples containing up to 10% DMSO and 10% DMF. Exposure to 100% DMSO is not recommended. Unfortunately, the compatibility of these devices with 100% DMF has not been determined.

      Helpful?

  3. Can I order the filter only, coming with no tube part?

    1 answer
    1. Unfortunately, no, this product is not available as a stand alone item. The tubes are available separately under the product UFC50VL96.

      Helpful?

  4. Is the 0.5 ml 3KDa Amicon device (UFC5003) compatible with a solution containing 5% SDS (Sodium Dodecyl Sulfate)?

    1 answer
    1. The Amicon® Ultra centrifugal devices are designed for processing biological fluids and aqueous solutions. Amicon® Ultra Filter Devices are compatible with concentrations of Sodium dodecyl sulfate (SDS)of 0.1% or less. Prior to use, it is important to verify the chemical compatibility of the sample with the device.

      Helpful?

  5. Hi, Do these filters come with expiry dates? If so where would I be able to find them on the product?

    1 answer
    1. This product is not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one year warranty from the date of shipment. For more information you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

  6. How long should I centrifugate at 14.000 g and how long for the reverse spin at 1000 g?

    1 answer
    1. Typical concentration time for the 3kD membrane device is approximately 30 minutes. However, spin times may need to be adjusted to either a shorter or longer time depending on the sample starting volume, initial concentration, composition, and desired minimum volume. The reverse spin is 1000g for 2 minutes. Please review the product user guide at the link below:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/271/340/pr05485-rev0219.pdf

      Helpful?

  7. Hi, how many g's can be used for the ultracell-3K #UFC500396 product?

    1 answer
    1. The recommended speed with a 35 degree fixed angle rotor is 14,000 g with a reverse spin of 1000 g. This information can be found at this link: https://www.sigmaaldrich.com/products/protein-biology/protein-sample-prep/amicon-centrifugals#ultracentrifugal

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service