MilliporeSigma
  • The novel small-molecule antagonist MMG-11 preferentially inhibits TLR2/1 signaling.
The novel small-molecule antagonist MMG-11 preferentially inhibits TLR2/1 signaling.
Biochemical pharmacology (2019-11-05)
Maria Grabowski, Manuela S Murgueitio, Marcel Bermudez, Gerhard Wolber, Günther Weindl
ABSTRACT
Toll-like receptor 2 (TLR2) forms heterodimers with either TLR1 or TLR6 to induce protective early inflammatory responses to pathogen- and damage-associated molecular patterns. However, excessive activation is associated with inflammatory and metabolic diseases. Several TLR2 antagonists have been described but pharmacological characterization is still at an early stage. Previously, we identified the potent and selective TLR2 antagonist MMG-11 by computational modelling and experimental validation. Here, we characterized the TLR2 antagonists MMG-11 and CU-CPT22 as well as the TIR-domain binding TLR2 antagonist C29 in TLR-overexpressing promoter cells as well as human and mouse macrophages. In line with our recent studies, MMG-11 abrogated pro-inflammatory cytokine secretion and NF-κB activation induced by different bacterial TLR2 agonists. MMG-11 preferentially inhibited TLR2/1 signaling in promoter cells stably expressing TLR2 heterodimers and mouse macrophages. Furthermore, the TLR2 antagonist blocked ligand-induced interaction of TLR2 with MyD88 and reduced MAP kinase and NF-κB activation. MMG-11 and CU-CPT22 but not C29 displaced Pam3CSK4 in an indirect binding assay confirming the competitive mode of action of MMG-11 and CU-CPT22. Isobologram analysis revealed additive and synergistic effects when the non-competitive antagonist C29 was combined with the competitive antagonist MMG-11 or CU-CPT22, respectively. In conclusion, we provide evidence that MMG-11 acts as a competitive antagonist with a predominance for the TLR2/1 heterodimer in human and mouse cells. Our results also indicate that MMG-11 is a model compound for studying TLR2 signaling.
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MMG-11, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
TLR2-IN-C29, ≥98% (HPLC)