Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2845

Sigma-Aldrich

Betrixaban

≥98% (HPLC)

Synonym(s):

MK-4448, MLN1021, N-(5-Chloro-2-pyridinyl)-2-[[4-[(dimethylamino)iminomethyl]benzoyl]amino]-5-methoxybenzamide, N-(5-Chloropyridin-2-yl)-2-(4-(N,N-dimethylcarbamimidoyl)benzamido)-5-methoxybenzamide, PRT-054021, PRT054021

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22ClN5O3
CAS Number:
Molecular Weight:
451.91
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CN=C(C=C1)NC(C2=C(NC(C3=CC=C(C(N(C)C)=N)C=C3)=O)C=CC(OC)=C2)=O

InChI

1S/C23H22ClN5O3/c1-29(2)21(25)14-4-6-15(7-5-14)22(30)27-19-10-9-17(32-3)12-18(19)23(31)28-20-11-8-16(24)13-26-20/h4-13,25H,1-3H3,(H,27,30)(H,26,28,31)

InChI key

XHOLNRLADUSQLD-UHFFFAOYSA-N

Biochem/physiol Actions

Betrixaban is an orally active, active site-targeting, highly potent and selective factor Xa (fXa) inhibitor (IC50 = 1.5 nM; Ki = 117 pM) with much reduced plasma kallikrein inhibitory activity (IC50 = 6.3 μM) and little potency against thrombin, trypsin, t-PA, aPC or plasmin inhibition (IC50 >10 μM). Betrixaban exhibits good pharmacokinetic properties and anticoagulant efficacy against various thromboembolism (VTE) events in vivo.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2

Target Organs

Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Menno V Huisman et al.
European heart journal supplements : journal of the European Society of Cardiology, 20(Suppl E), E12-E15 (2018-07-07)
Venous thromboembolism (VTE) in acute medically ill patients is a leading cause of in-hospital morbidity and mortality. A majority of these VTE events occur post-discharge, and patients remain at increased VTE risk for up to 3 months post-discharge. Recent clinical trials
Penglie Zhang et al.
Bioorganic & medicinal chemistry letters, 19(8), 2179-2185 (2009-03-20)
Systematic SAR studies of in vitro factor Xa inhibitory activity around compound 1 were performed by modifying each of the three phenyl rings. A class of highly potent, selective, efficacious and orally bioavailable direct factor Xa inhibitors was discovered. These
Romain Siriez et al.
Thrombosis and haemostasis, 118(7), 1203-1214 (2018-06-12)
Betrixaban is a novel direct oral factor Xa inhibitor approved by the Food and Drug Administration for prophylaxis of venous thromboembolism in adult patients hospitalized for an acute illness at risk for thromboembolic complications. Assessment of the anti-coagulant effect of
Emmanuel J Favaloro et al.
Seminars in thrombosis and hemostasis, 41(2), 208-227 (2015-02-24)
A new generation of antithrombotic agents has recently emerged. These provide direct inhibition of either thrombin (factor IIa [FIIa]) or FXa, and are increasingly replacing the classical anticoagulants (heparin and coumarins such as warfarin) in clinical practice for a variety
Meyer Michel Samama et al.
Clinics in laboratory medicine, 34(3), 503-517 (2014-08-30)
New oral factor Xa inhibitors are intended to progressively substitute the oral vitamin K antagonists and parenteral indirect inhibitors of factor Xa in the prevention and treatment of venous and arterial thromboembolic episodes. This article focuses on the main clinical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service