Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

422175

Sigma-Aldrich

L-165,041

A cell-permeable phenoxyacetic acid derivative that acts as a potent and selective peroxisome proliferator activator receptor δ (PPARδ) agonist (Ki = 6 nM for hPPARδ and 730 nM for hPPARγ).

Synonym(s):

L-165,041, Compound P, L165041, 4-[3-(2-Propyl-3-hydroxy-4-acetyl)phenoxy]propyloxyphenoxy-acetic acid, PPAR Agonist VIII, PPARα Agonist III, PPARγ Agonist VII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26O7
CAS Number:
Molecular Weight:
402.44
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

1 M NaOH: 40 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O(CCCOc2ccc(cc2)OCC(=O)O)c1c(c(c(cc1)C(=O)C)O)CCC

InChI

1S/C22H26O7/c1-3-5-19-20(11-10-18(15(2)23)22(19)26)28-13-4-12-27-16-6-8-17(9-7-16)29-14-21(24)25/h6-11,26H,3-5,12-14H2,1-2H3,(H,24,25)

InChI key

HBBVCKCCQCQCTJ-UHFFFAOYSA-N

General description

A cell-permeable phenoxyacetic acid derivative that acts as a potent and selective peroxisome proliferator activator receptor δ (PPARδ) agonist (Ki = 6 nM for hPPAR δ and 730 nM for hPPARγ). Induces adipocyte differentiation in NIH-PPARδ cells at 500 nM. Also reported to raise total cholesterol in insulin-resistant db/db mice without altering glucose or triglyceride levels. Shown to increase uncoupling protein 3 (UCP3) gene expression in L6 myotubes. Also shown to inhibit cytokine-induced expression of vascular cell adhesion molecule-1 (VCAM-1) and the secretion of monocyte chemotactic protein-1 (MCP-1) in Eahy926 cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Peroxisome proliferator activator receptor δ (PPAR@delta;)
Product does not compete with ATP.
Reversible: no
Target Ki: 6 nM for hPPARδ and 730 nM for hPPARγ

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Rival, Y., et al. 2002. Eur. J. Pharmacol.435, 143.
Castrillo, A., et al. 2001. J. Biol. Chem.276, 34082.
Hansen, J.B., et al. 2001. J. Biol. Chem.276, 3175.
Son, C., et al. 2001. Endocrinology142, 4189.
Wilkie, N., et al. 2001. J. Neurochem.78, 1135.
Leibowitz, M.D., et al. 2000. FEBS Lett.473, 333.
Berger, J., et al. 1999. J. Biol. Chem.274, 6718.
Lim, H., et al. 1999. Genes Dev.13, 1561.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service