Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1533

Sigma-Aldrich

(±)-Marinopyrrole A

≥95% (HPLC)

Synonym(s):

(±)-Maritoclax, 1,1′-(4,4′,5,5′-Tetrachloro[1,3′-bi-1H-pyrrole]-2,2′-diyl)bis[1-(2-hydroxyphenyl)-methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12Cl4N2O4
CAS Number:
Molecular Weight:
510.15
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

light yellow to dark yellow

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

Clc1[n](c(cc1Cl)C(=O)c4c(cccc4)O)c2c([nH]c(c2Cl)Cl)C(=O)c3c(cccc3)O

InChI

1S/C22H12Cl4N2O4/c23-12-9-13(19(31)10-5-1-3-7-14(10)29)28(22(12)26)18-16(24)21(25)27-17(18)20(32)11-6-2-4-8-15(11)30/h1-9,27,29-30H

InChI key

QYPJBTMRYKRTFG-UHFFFAOYSA-N

General description

Marinopyrrole A (Maritoclax) is a member of phenolic 1,3′-bipyrroles family targeting actin group.

Biochem/physiol Actions

Marinopyrrole A (Maritoclax) is a potent and selective inhibitor of Bcl-2 family member Mcl-1, originally isolated from marine streptomycetes for its antimicrobial activity against methicillin-resistant Staphylococcus aureus. Marinopyrrole A directly binds to Mcl-1 and targets it for proteasomal degradation. Marinopyrrole A induced apoptosis in Mcl-1-dependent cancer cells and sensitized cancer cells to ABT-737.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chambers C Hughes et al.
Journal of the American Chemical Society, 131(34), 12094-12096 (2009-08-14)
The targeting of marinopyrrole A to actin was identified using a fluorescent dye transfer strategy. The process began by appending a carboxylic acid terminal tag to a phenol in the natural product. The resulting probe was then studied in live

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service