Skip to Content
MilliporeSigma

1.60338

Cerium(IV) sulfate solution

c(Ce(SO₄)2 * 4 H₂O) = 0.1 mol/l (0.1 N), reag. USP, ready-to-use volumetric solution for titration, Titripur®

Synonym(s):

Ceric sulfate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Ce(SO4)2
CAS Number:
Molecular Weight:
332.24
NACRES:
NA.21
UNSPSC Code:
12352302
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Cerium(IV) sulfate solution, c(Ce(SO4)2 * 4 H2O) = 0.1 mol/l (0.1 N) (20° C), Titripur®, reag. USP

Quality Segment

agency

reag. USP, traceable to NIST

product line

Titripur®

form

liquid

quality

Analyzed in our ISO 17025 accredited QC lab

reaction suitability

reaction type: Redox Reactions

concentration

0.0995 - 0.1005 mol (/L)

measuring range

±0.0003 (mol/L)

technique(s)

titration: suitable

suitability

suitable for (Traceability NIST SRM)

storage temp.

room temp

SMILES string

[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Ce.2H2O4S/c;2*1-5(2,3)4/h;2*(H2,1,2,3,4)/q+4;;/p-4

InChI key

VZDYWEUILIUIDF-UHFFFAOYSA-J

Features and Benefits

Digital Titration

ChemiSphere™ App for Digital Data Transfer to OMNIS Titration Instruments from Metrohm

Discover the ChemiSphere™ App for secure, rapid data transfer to OMNIS titrators, enhancing efficiency in the titration workflow with Titripur® and Aquastar® reagents. ChemiSphere App for Digital Reagent Data Transfer



SmartChemicals for Digital Data Transfer to Mettler Toledo Titration Instruments

Explore digital data transfer with our SmartChemicals for Mettler Toledo titrators—fast, easy, and safe, accelerating the titration process with Titripur®, Aquastar® reagents, and Certipur® and Aquastar® standards. Supelco® SmartChemicals for Titration
Accreditation:
This volumetric solution is analyzed by our calibration laboratory D-K-15185-01-00 which is accredited according to DIN EN ISO/IEC 17025 for analysis of amount-of-substance concentrations in volumetric solutions by DAkkS (Deutsche Akkreditierungsstelle - German National Accreditation Body). The accreditation certificate can be found at www.sigmaaldrich.com/ISO17025.

Analysis Note

Form liquid
Amount-of-substance concentration 0.0995 - 0.1005 mol/L
Measurement uncertainty ± 0.0003 mol/L
Traceability NIST SRM

The concentration is determined by volumetric titration and refers to 20°C.
The amount-of-substance concentration of this volumetric solution is traceable to a primary standard reference material (SRM) from the National Institute of Standards and Technology, Gaithersburg, USA (NIST SRM 8040 di-sodium oxalate) by means of volumetric standard di-sodium oxalate (article number 1.02407), certified reference material according to ISO 17034, analyzed by our accredited calibration laboratory of Merck KGaA, Darmstadt, Germany according to DIN EN ISO/IEC 17025. The uncertainty is expressed as expanded measurement uncertainty with a coverage factor k=2 covering a confidence level of 95%.

Note: The titer is a correction factor to correct for variations of the volumetric solution, the titration equipment, the temperature and other laboratory conditions. For correct titration results it is recommended to determine a titer with the laboratory specific equipment and under laboratory specific conditions directly after opening a new bottle and at regular time intervals.

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
ChemiSphere is a trademark of Merch KGaA, Darmstadt, Germany
Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany
Titripur is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Cerium(IV) sulfate solution


Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library