Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0805

Sigma-Aldrich

CID16020046

≥98% (HPLC)

Synonym(s):

4-[4,6-Dihydro-4-(3-hydroxyphenyl)-3-(4-methylphenyl)-6-oxopyrrolo[3,4-c]pyrazol-5(1H)-yl]-benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H19N3O4
CAS Number:
Molecular Weight:
425.44
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[nH]1nc(c3c1C(=O)N(C3c5cc(ccc5)O)c4ccc(cc4)C(=O)O)c2ccc(cc2)C

InChI

1S/C25H19N3O4/c1-14-5-7-15(8-6-14)21-20-22(27-26-21)24(30)28(18-11-9-16(10-12-18)25(31)32)23(20)17-3-2-4-19(29)13-17/h2-13,23,29H,1H3,(H,26,27)(H,31,32)

InChI key

VGUQVYZXABOXCX-UHFFFAOYSA-N

Application

CID16020046 has been used as an antagonist of G-protein coupled receptor (GPR55) to test its effect on nociceptive behavior in rats induced with formalin.

Biochem/physiol Actions

CID16020046 is a potent and a selective GPR55 antagonist that inhibits GPR55-mediated ERK1/2 phosphorylation. CID16020046 inhibits LPI-induced Ca2+ signaling in HEK-GPR55 cells.
CID16020046 is a potent and a selective GPR55 antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bright N Okine et al.
Neuroscience, 443, 19-29 (2020-07-17)
The G-protein coupled receptor, GPR55, modulates nociceptive processing. Given the expression of GPR55 in the anterior cingulate cortex (ACC), a key brain region involved in the cognitive and affective dimensions of pain, the present study tested the hypothesis that GPR55

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service