Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3585

Sigma-Aldrich

PSB-CB5

≥98% (HPLC)

Synonym(s):

5H-Imidazo[2,1-b][1,3]thiazin-3(2H)-one, 2-[[3-[(4-chlorophenyl)methoxy]phenyl]methylene]-6,7-dihydro-, (2Z)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17ClN2O2S
CAS Number:
Molecular Weight:
384.88
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

O=C1N2C(SCCC2)=N/C1=C\C3=CC(OCC4=CC=C(C=C4)Cl)=CC=C3

InChI key

XJBQRMOGMULGPP-PDGQHHTCSA-N

Biochem/physiol Actions

PSB-CB5 is a potent and selective GPR18 antagonist that inhibits Δ9-THC-induced GPR18 β-arrestin recruitment (IC50 = 279 nM; [THC] = EC50 = 7.5 μM) with no potency against human GPR55 (IC50 >10 μM) and little or no affinity toward human CB2 and CB1 (Ki = 4.03 μM & >10 μM, respectively).
Potent and selective GPR18 antagonist with little or no potency towrd human GPR55, CB1 or CB2.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bicyclic imidazole-4-one derivatives: a new class of antagonists for the orphan G protein-coupled receptors GPR18 and GPR55
MedChemComm, 5(5), 632-649 (2014)
Effects of GPR18 Ligands on Body Weight and Metabolic Parameters in a Female Rat Model of Excessive Eating
Pharmaceuticals (Basel, Switzerland), 14(3), 270-270 (2021)
Douglas J Hermes et al.
Experimental neurology, 341, 113699-113699 (2021-03-20)
Human immunodeficiency virus type 1 (HIV-1) is known to provoke microglial immune responses which likely play a paramount role in the development of chronic neuroinflammatory conditions and neuronal damage related to HIV-1 associated neurocognitive disorders (HAND). In particular, HIV-1 Tat

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service