Skip to Content
MilliporeSigma
All Photos(1)

Documents

SMB00315

Sigma-Aldrich

Rosarin

≥98% (HPLC)

Synonym(s):

Rosarin, (2E)-3-Phenyl-2-propen-1-yl 6-O-a-L-arabinofuranosyl-β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O10
CAS Number:
Molecular Weight:
428.43
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

biological source

plant (Rhodiola rosea)

Quality Level

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](O)[C@H](OC/C=C/C2=CC=CC=C2)O[C@H](CO[C@H]3[C@H](O)[C@@H](O)[C@H](CO)O3)[C@H]1O

InChI

1S/C20H28O10/c21-9-12-14(22)17(25)20(29-12)28-10-13-15(23)16(24)18(26)19(30-13)27-8-4-7-11-5-2-1-3-6-11/h1-7,12-26H,8-10H2/b7-4+/t12-,13+,14-,15+,16-,17+,18+,19+,20+/m0/s1

InChI key

IEBFEMIXXHIISM-YZOUKVLTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rosarin is a cinnamyl alcohol glycoside and one of the main biologically active compounds found in Rhodiola rosea L. (golden root).
Rosarin is a cinnamyl alcohol glycoside found in Rhodiola rosea.

Biochem/physiol Actions

Rosarin has been shown to reduce fatigue, improve exercise endurance, and increase mental performance.
Rosarin is known to alter the TNF-related apoptosis-inducing ligand (TRAIL) expression by modulating extracellular signal-regulated kinase (ERK) signaling.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Does Rhodiola rosea possess ergogenic properties?
Walker, et al.
International Journal of Sport Nutrition and Exercise Metabolism, 16(3) (2006)
Production of Plant Derived Natural Compounds Through Hairy Root Culture (2017)
Liquid chromatographic method for the quantification of salidroside and cinnamyl alcohol glycosides for quality control of golden root (Rhodiola rosea L.)
Ajdert P, et al.,
Journal of Applied Research on Medicinal and Aromatic Plants, 26, 100364-100364 (2022)
Andrey S Marchev et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 108(Pt B), 419-428 (2017-02-13)
Rhodiola rosea L. extracts have shown neuroprotective, anti-fatigue, anti-inflammatory and anti-tumor properties. However, the studies on their effect on T cell function are rather scarce. We examined the potential of R. rosea extract and its major constituents - salidroside, rosarin, rosavin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service