Skip to Content
MilliporeSigma

SML0336

Rosavin

≥98% (HPLC), Antioxidant, powder

Synonym(s):

(2E)-3-Phenylprop-2-en-1-yl 6-O-α-L-arabinopyranosyl-β-D-glucopyranoside, (2R,3R,4S,5S,6R)-2-[(E)-3-Phenylprop-2-enoxy]-6-([(2S,3R,4S,5S)-3,4,5-trihydroxyoxan-2-yl]oxymethyl)oxane-3,4,5-triol, (E)-3-Phenyl-2-propenyl 6-O-a-L-arabinopyranosyl-β-D-glucopyranoside, Rosavidin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H28O10
CAS Number:
Molecular Weight:
428.43
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Rosavin, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -47 to -57° (C=1, MeOH CHCl3 (1:1))

color

white to beige

solubility

H2O: ≥5 mg/mL (warmed)

storage temp.

−20°C

SMILES string

O[C@H]1CO[C@@H](OC[C@H]2O[C@@H](OC\C=C\c3ccccc3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H]1O

InChI

1S/C20H28O10/c21-12-9-28-19(17(25)14(12)22)29-10-13-15(23)16(24)18(26)20(30-13)27-8-4-7-11-5-2-1-3-6-11/h1-7,12-26H,8-10H2/b7-4+/t12-,13+,14-,15+,16-,17+,18+,19-,20+/m0/s1

InChI key

RINHYCZCUGCZAJ-IPXOVKFZSA-N

Biochem/physiol Actions

Rosavin is a well-known glycoside antioxidant from Rhadiola rosea (Golden Root, or Arctic Root) that exhibits antidepressant, anxiolytic and adaptogenic properties.
Rosavin is an antidepressant and anxiolytic; antiapoptotic.
Rosavin is a benzyl propylene based glycoside and is used in treating pulmonary fibrosis. Rosavin elicits inhibition of expression of tumor necrosis factor based apoptosis in Jurkat T cells.


Still not finding the right product?

Explore all of our products under Rosavin


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library