Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

60895

Sigma-Aldrich

Gum rosin

natural resin

Synonym(s):

Colophony, Rosin, gum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
11121502
NACRES:
NA.22

form

solid

acid value

150‑170

solubility

chloroform: 0.1 g/mL, clear, slightly yellow to deep yellow

InChI

1S/C20H30O2/c1-13(2)14-6-8-16-15(12-14)7-9-17-19(16,3)10-5-11-20(17,4)18(21)22/h7,12-13,16-17H,5-6,8-11H2,1-4H3,(H,21,22)/t16-,17+,19+,20+/m0/s1

InChI key

RSWGJHLUYNHPMX-ONCXSQPRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gum rosin is an exudate gum, which is extracted from pine trees. It consists of abiatic acid, palaustric acid and neoabetic acid.

Application

Gum rosin has been used:
  • for the preparation of testosome patches
  • to study its influence on the physical stability of concentrated oil in water emulsions
  • for the preparation of rosin nanoparticles containing hydrocortisone (HC)

Analysis Note

powder to solified mass or chunks, yellow to deep yellow

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

370.4 °F - closed cup

Flash Point(C)

188 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rosin nanoparticles as a drug delivery carrier for the controlled release of hydrocortisone
Lee CM, et al
Biotechnology Letters, 27(19), 1487-1490 (2005)
Ethosomes?novel vesicular carriers for enhanced delivery: characterization and skin penetration properties
Touitou E, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 65(3), 403-418 (2000)
Development of eco-friendly submicron emulsions stabilized by a bio-derived gum
Perez-Mosqueda LM, et al.
Colloids and Surfaces. B, Biointerfaces, 123, 797-802 (2014)
Ambra Masuzzo et al.
eLife, 8 (2019-10-30)
When facing microbes, animals engage in behaviors that lower the impact of the infection. We previously demonstrated that internal sensing of bacterial peptidoglycan reduces Drosophila female oviposition via NF-κB pathway activation in some neurons (Kurz et al., 2017). Although we
Contact allergy to a permanent marker.
Paloma Sánchez-Pedreño et al.
Dermatitis : contact, atopic, occupational, drug, 23(2), 96-97 (2012-06-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service